L-Tyrosine disodium salt dihydrate 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(S-3-(4-Hydroxyphenyl)alanine disodium salt dihydrate, Disodium 2-Amino-3-(4-oxidophenyl) propanoate dihydrate, L-3-(4-Hydroxyphenyl)alanine disodium salt dihydrate
J61770-22
J61770-36
J61770-A1
CAAAJ61770-22EA
269.34
CAD
CAAAJ61770-22
CAAAJ61770-36
CAAAJ61770-A1
L-Tyrosine disodium salt dihydrate 99%
L(-)-Tyrosine disodium salt
Powder
Purity: 99%
CAS No.: 122666-87-9
Molecular Formula: C9H9NNa2O3.2H2O
Formula Weight: 265.47 (225.15anhy)
Physical Form: Powder
Sensitivity: Hygroscopic
Appearance: White to pale brown
Optical Rotation: -13° (c=4 in water)
MDL No.: MFCD03840558
Formula:
C₉H₉NO₃Na₂·2H₂O MW: 261.9 Storage Temperature: Ambient |
MDL Number:
MFCD03840558 CAS Number: 122666-87-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...