Polymyxin B sulphate, Cell Culture grade Cell Culture
Supplier: Thermo Scientific Chemicals
|
Warning
|
J61763-03
J61763-06
CAAAJ61763-03EA
244.77
CAD
CAAAJ61763-03
CAAAJ61763-06
Polymyxin B sulphate, Cell Culture grade Cell Culture
Polymyxin B sulphate
Antibiotic for Gram-negative bacteria
White to pale yellow powder
Soluble in water
Formula:
C₅₅H₉₆N₁₆O₁₃·2H₂SO₄ MW: 1385.61 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00131991 CAS Number: 1405-20-5 EINECS: 215-774-7 Merck Index: 14,7573 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...