Acarbose 95%

Supplier: Thermo Scientific Chemicals

J61737-03 J61737-04 J61737-06
CAAAJ61737-03EA 354.99 CAD
CAAAJ61737-03 CAAAJ61737-04 CAAAJ61737-06
Acarbose 95%
Acarbose

A reversible alpha-glucosidase inhibitor


Powder


Applications: A reversible alpha-glucosidase inhibitor

Formula: C₂₅H₄₃NO₁₈
Storage Temperature: Ambient
MDL Number: MFCD00869592
CAS Number: 56180-94-0
EINECS: 260-030-7
Merck Index: 14,18

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR