Phosphate, 0.5 M Buffer Solution, pH 8.0, Thermo Scientific

Supplier: Thermo Scientific Chemicals

J61722-AK J61722-AP
CAAAJ61722-AKEA 200.11 CAD
CAAAJ61722-AK CAAAJ61722-AP
Phosphate, 0.5 M Buffer Solution, pH 8.0, Thermo Scientific
Buffers Biological Buffers

Liquid


CAS No.: 7558-80-7
Physical Form: Liquid
MDL No.: MFCD00003527

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR