Prazosin hydrochloride

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-(4-Amino-6,7-dimethoxy-2-quinazolinyl)-4-(2-furanylcarbonyl)piperazine Hydrochloride, (4-(4-Amino-6,7-dimethoxyquinazolin-2-yl)piperazin-1-yl)(furan-2-yl)methanone hydrochloride, Prazosin HCl

J61712-03 J61712-MD
CAAAJ61712-03EA 755.82 CAD
CAAAJ61712-03 CAAAJ61712-MD
Prazosin hydrochloride
Prazosin hydrochloride

An alpha adrenoceptor antagonist


Powder


Applications: An alpha adrenoceptor antagonist

Formula: C₁₉H₂₁N₅O₄
MW: 419.87 g/mol
Melting Pt: 285 °C
Storage Temperature: Ambient
MDL Number: MFCD00058177
CAS Number: 19237-84-4
EINECS: 242-903-4
Merck Index: 14,7717

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR