Tyrphostin B46 ≥98%

Supplier: Thermo Scientific Chemicals

Synonyms: (E)-2-Cyano-3-(3,4-dihydroxyphenyl)-N-(3-phenylpropyl)prop-2-enamide, (E)-2-Cyano-3-(3,4-dihydroxyphenyl)-N-(3-phenylpropyl)-2-propenamide, AG 555, (E)-2-Cyano-3-(3,4-dihydroxyphenyl)-N-(3-phenylpropyl)acrylamide, Tyrphostin AG-555

J61704-MA J61704-MF
CAAAJ61704-MAEA 544.8 CAD
CAAAJ61704-MA CAAAJ61704-MF
Tyrphostin B46 ≥98%
Tyrphostin B46

Inhibitor of EGF receptor kinase activity


Powder


Applications: Inhibitor of EGF receptor kinase activity

Formula: C₁₉H₁₈N₂O₃
Storage Temperature: Refrigerator
MDL Number: MFCD00209865
CAS Number: 133550-34-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR