H-7 dihydrochloride

Supplier: Thermo Scientific Chemicals

J61697-MA J61697-MF
CAAAJ61697-MAEA 518.87 CAD
CAAAJ61697-MA CAAAJ61697-MF
H-7 dihydrochloride
H-7 dihydrochloride

Protein kinase inhibitor. Inhibits cyclic nucleotide dependent protein kinase (PKA) and protein kinase C


Powder

Formula: C₁₄H₁₇N₃O₂S·2HCl
Melting Pt: 191…193 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00036961
CAS Number: 108930-17-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR