Lecithin (from soy bean) ≥90%
Supplier: Thermo Scientific Chemicals
J61675-18
J61675-22
J61675-30
CAAAJ61675-18EA
39.15
CAD
CAAAJ61675-18
CAAAJ61675-22
CAAAJ61675-30
Lecithin (from soy bean) ≥90%
Lecithin
Naturally occuring lipid that protects cells from oxidation. Useful as an emulsifier
- Pale brown to yellow Solid
- Odourless
- Beilstein Reference: 5209585
MDL Number:
MFCD00147406 CAS Number: 8002-43-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...