Guanosine-5'-diphosphate disodium salt
Supplier: Thermo Scientific Chemicals
Synonyms:
((2R,3S,4R,5R)-5-(2-Amino-6-oxo-3,6-dihydro-9H-purin-9-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl trihydrogen diphosphate, disodium salt, Guanosine 5'-diphosphate disodium salt
J61646-MC
J61646-ME
CAAAJ61646-MCEA
259.36
CAD
CAAAJ61646-MC
CAAAJ61646-ME
Guanosine-5'-diphosphate disodium salt
Guanosine 5'-diphosphate disodium salt
Powder
CAS No.: 7415-69-2
Molecular Formula: C10H13N5Na2O11P2
Formula Weight: 487.17
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: White
MDL No.: MFCD00084665
Formula:
C₁₀H₁₃N₅Na₂O₁₁P₂ MW: 487.17 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00070121 CAS Number: 7415-69-2 EINECS: 231-026-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...