Adenosine 5'-monophosphate disodium salt (AMP disodium salt) hydrate

Supplier: Thermo Scientific Chemicals

Synonyms: 5′-AMP-Na2 hydrate

J61643-06 J61643-14
CAAAJ61643-06EA 63.11 CAD
CAAAJ61643-06 CAAAJ61643-14 CAAAJ61643-22
Adenosine 5'-monophosphate disodium salt (AMP disodium salt) hydrate
Adenosine 5'-monophosphate disodium salt (AMP disodium salt) hydrate

Powder


CAS No.: 4578-31-8 Molecular Formula: C10H12N5NaO7P Formula Weight: 391.20 Storage Temperature: -30°C to -10°C Physical Form: Powder Appearance: White Beilstein Reference: 3586732 MDL No.: MFCD00065023

Formula: C₁₀H₁₂N₅Na₂O₇P·H₂O
MW: 391.2 g/mol (anhy)
Storage Temperature: Refrigerator
MDL Number: MFCD00065023
CAS Number: 4578-31-8
EINECS: 224-961-2
Merck Index: 14,158

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR