β-Dihydronicotinamide adenine dinucleotide disodium salt (NADH-Na₂, reduced form) 97%
Supplier: Thermo Scientific Chemicals
Synonyms:
Nicotinamide adenine dinucleotide reduced, disodium salt (reduced form), NADH disodium salt, ((2R,3S,4R,5R)-5-(6-Aminopurin-9-yl)-3,4-dihydroxy-oxolan-2-yl)methoxy-((((2R,3S,4R,5R)-5-(3-carbamoyl-4H-pyridin-1-yl)-3,4-dihydroxy-oxolan-2-yl)methoxy)hydroxyphosphoryl)oxyphosphinic acid disodium salt
Powder
Purity: 98%
CAS No.: 606-68-8
Molecular Formula: C21H27N7Na2O14P2
Formula Weight: 709.41
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Beilstein Reference: 5230241
MDL No.: MFCD00036200
Formula:
C₂₁H₂₇N₇Na₂O₁₄P₂ MW: 709.41 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00036200 CAS Number: 606-68-8 EINECS: 210-123-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...