Erbstatin analog
Supplier: Thermo Scientific Chemicals
|
Danger
|
J61612-LB0
J61612-MA
CAAAJ61612-LB0EA
201.73
CAD
CAAAJ61612-LB0
CAAAJ61612-MA
CAAAJ61612-MCR
Erbstatin analog
Erbstatin analog
An inhibitor of the EGF receptor associated tyrosine kinase
CAS No.: 63177-57-1
Molecular Formula: C10H10O4
Formula Weight: 194.19
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: Yellow
Solubility: Soluble in methanol or 0.1M NaOH. Insoluble in water
UN No.: 2811
MDL No.: MFCD00132932
An inhibitor of the EGF receptor associated tyrosine kinase
Formula:
C₁₀H₁₀O₄ Storage Temperature: Freezer |
MDL Number:
MFCD00132932 CAS Number: 63177-57-1 UN: 2811 ADR: "6.1",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...