Ergocalciferol (Vitamin D2, Calciferol)
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
(S,Z)-3-((E)-2-((1R,3aS,7aR)-1-((2R,5R,E)-5,6-Dimethylhept-3-en-2-yl)-7a-methylhexahydro-1H-inden-4(2H)-ylidene)ethylidene)-4-methylenecyclohexanol
J61610-03
J61610-09
CAAAJ61610-03EA
59.41
CAD
CAAAJ61610-03
CAAAJ61610-09
Ergocalciferol (Vitamin D2, Calciferol)
Ergocalciferol (Vitamin D2, Calciferol)
Powder
CAS No.: 50-14-6
Molecular Formula: C28H44O
Formula Weight: 396.65
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White
Melting Point: 114-118°
Beilstein Reference: 1916682
Merck Reference: 14,10018
UN No.: 2811
MDL No.: MFCD00166988
Formula:
C₂₈H₄₄O MW: 396.66 g/mol Melting Pt: 114…118 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00166988 CAS Number: 50-14-6 EINECS: 200-014-9 UN: 2811 ADR: 6.1 III Merck Index: 14,10018 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...