Streptozocin 97+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Methyl-1-nitroso-3-((2S,3R,4R,5S,6R)-2,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-3-yl)urea, Streptozotocin

J61601-ME J61601-03
CAAAJ61601-MEEA 238.58 CAD
CAAAJ61601-ME CAAAJ61601-03
Streptozocin 97+%
Streptozocin

Antibiotic and antitumor agent. Potent methylating agent for DNA


Purity: 97+% CAS No.: 18883-66-4 Molecular Formula: C8H15N3O7 Formula Weight: 265.22 Storage Temperature: -30°C to -10°C Physical Form: Powder Sensitivity: Hygroscopic Appearance: White to Off-white Melting Point: 121° Solubility: Soluble in water or 100% ethanol Beilstein Reference: 2060675 Merck Reference: 14,8832 MDL No.: MFCD00006607


Antibiotic and antitumor agent. Potent methylating agent for DNA

Formula: C₈H₁₅N₃O₇
MW: 265.2 g/mol
Melting Pt: 121 °C
Storage Temperature: Freezer
MDL Number: MFCD00006607
CAS Number: 18883-66-4
EINECS: 242-646-8
Merck Index: 14,8832

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR