Emodin

Supplier: Thermo Scientific Chemicals
Warning

J61600-03 J61600-MC J61600-MD
CAAAJ61600-03EA 1611.65 CAD
CAAAJ61600-03 CAAAJ61600-MC CAAAJ61600-MD
Emodin
Emodin

A protein tyrosine kinase inhibitor


Powder


Applications: A protein tyrosine kinase inhibitor

Formula: C₁₅H₁₀O₅
Storage Temperature: Refrigerator
MDL Number: MFCD00001207
CAS Number: 518-82-1
EINECS: 208-258-8
Merck Index: 14,3561

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR