Quisqualic Acid ≥99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: L-Quisqualic acid, (S)-2-Amino-3-(3,5-dioxo-1,2,4-oxadiazolidin-2-yl)propanoic acid

J61591-LB0 J61591-MA
CAAAJ61591-LB0EA 389.47 CAD
CAAAJ61591-LB0 CAAAJ61591-MA CAAAJ61591-MF
Quisqualic Acid ≥99%
Quisqualic acid

Group I mGlu receptor agonist. AMPA agonist


Group I mGlu receptor agonist. AMPA agonist


Group I mGlu receptor agonist. AMPA agonist

Formula: C₅H₇N₃O₅
Melting Pt: 185…187 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00069337
CAS Number: 52809-07-1
Merck Index: 14,8085

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR