Dicloxacillin sodium salt monohydrate

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 3-(2,6-Dichlorophenyl)-5-methyl-4-isoxazolyl penicillin sodium salt monohydrate, Sodium (2S,5R,6R)-6-(3-(2,6-dichlorophenyl)-5-methylisoxazole-4-carboxamido)-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate hydrate, Dicloxacillin Sodium hydrate

J61581-03 J61581-06
CAAAJ61581-03EA 180.25 CAD
CAAAJ61581-03 CAAAJ61581-06
Dicloxacillin sodium salt monohydrate
Dicloxacillin sodium salt monohydrate

An antibiotic used in the treatment of staphyloccoci bacteria that are resistant to penicillin G


Powder


Applications: An antibiotic used in the treatment of staphyloccoci bacteria that are resistant to penicillin G

Formula: C₁₉H₁₆Cl₂N₃NaO₅S·1H₂O
MW: 510.33 g/mol
Melting Pt: 222…225 °C
Storage Temperature: Ambient
MDL Number: MFCD00056865
CAS Number: 13412-64-1
EINECS: 603-794-2
Merck Index: 14,3084

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR