Potassium Phosphate, 0.5 M Buffer Solution, pH 7.5, Thermo Scientific
Supplier: Thermo Scientific Chemicals
J61553-AK
J61553-AP
CAAAJ61553-AKEA
135.99
CAD
CAAAJ61553-AK
CAAAJ61553-AP
Potassium Phosphate, 0.5 M Buffer Solution, pH 7.5, Thermo Scientific
Buffers
Biological Buffers
Liquid
CAS No.: 7778-77-0
Physical Form: Liquid
MDL No.: MFCD00011401
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...