Tween 20 Blocking Buffer, 1% in PBS (10X), Thermo Scientific
Supplier: Thermo Scientific Chemicals
J61544-AP
J61544-K2
J61544-K3
CAAAJ61544-APEA
121.41
CAD
CAAAJ61544-AP
CAAAJ61544-K2
CAAAJ61544-K3
Tween 20 Blocking Buffer, 1% in PBS (10X), Thermo Scientific
Buffers
Electrophoresis Buffers
Blocking Buffers
Viscous liquid
Storage Temperature: 2°C to 8°C
Physical Form: Viscous liquid
Appearance: Clear
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...