Acetyl-L-carnitine hydrochloride

Supplier: Thermo Scientific Chemicals
Warning

J61536-06 J61536-14
CAAAJ61536-06EA 256.18 CAD
CAAAJ61536-06 CAAAJ61536-14
Acetyl-L-carnitine hydrochloride
Acetyl-L-carnitine hydrochloride

Weak cholinergic agonist


White crystalline powder

Formula: C₉H₁₇NO₄
Storage Temperature: Refrigerator
MDL Number: MFCD00082230
CAS Number: 5080-50-2
Merck Index: 14,84

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR