Cyclopamine 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (2'R,3S,3'R,3a'S,6aS,6bS,6'S,7a'R,11aS,11bR)-3',6',10,11b-tetramethyl-1,2,3,3a',4,4',5',6,6a,6b,6',7,7',7a',8,11,11a,11b-octadecahydro-3'H-spiro[benzo[a]fluorene-9,2'-furo[3,2-b]pyridin]-3-ol

J61528-MB J61528-MF
CAAAJ61528-MBEA 833.5 CAD
CAAAJ61528-MB CAAAJ61528-MF
Cyclopamine 99%
Cyclopamine

Inhibits hedgehog/smoothened signaling. A steroidal alkaloid


Powder


Applications: Inhibits hedgehog/smoothened signaling. A steroidal alkaloid

Formula: C₂₇H₄₁NO₂
MW: 411.63 g/mol
Melting Pt: 234…237 °C
Storage Temperature: Freezer
MDL Number: MFCD09878269
CAS Number: 4449-51-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR