Aspartame
Supplier: Thermo Scientific Chemicals
J61523-06
J61523-14
J61523-22
CAAAJ61523-06EA
148.8
CAD
CAAAJ61523-06
CAAAJ61523-14
CAAAJ61523-22
Aspartame
Proteins and Peptides
A non-nutritive sweetener and dipeptide ester about 160 times sweeter than sucrose.
- EINECS: 245-261-3
- Melting point: 243 to 245 °C
- Merck Index: 14,839
Purity: 98%
CAS No.: 22839-47-0
Molecular Formula: C14H18N2O5
Formula Weight: 294.30
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White
Melting Point: 243-245°
Beilstein Reference: 2223850
MDL No.: MFCD00002724
Applications: A non-nutritive sweetener and dipeptide ester about 160 times sweeter than sucrose
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...