Methyl green zinc chloride salt ≥98%, ee 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
J61509-14
J61509-22
CAAAJ61509-14EA
283.23
CAD
CAAAJ61509-14
CAAAJ61509-22
Methyl green zinc chloride salt ≥98%, ee 98%
Methyl Green zinc chloride salt
DNA stain useful as tracking dye in acid buffers for electrophoresis. Binds selectively to AT-rich regions of native DNA.
Powder
DNA stain useful as tracking dye in acid buffers for electrophoresis. Binds selectively to AT-rich regions of native DNA.
Formula:
C₂₇H₃₅BrClN₃·xZnCl₂ Melting Pt: >300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00151094 CAS Number: 7114-03-6 EINECS: 230-415-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...