Neomycin trisulfate salt hydrate
Supplier: Thermo Scientific Chemicals
|
Danger
|
An antibiotic. Acts as an inhibitor of protein biosynthesis
Neomycin sulfate is an aminoglycoside antibiotic and calcium channel protein inhibitor. Neomycin sulfate also binds to prokaryotic ribosomes inhibiting translation and is effective against gram-positive and gram-negative bacteria.
Neomycin sulfate inhibits Phospholipase C via binding to inositol phospholipids. It also inhibits phosphatidylcholine-PLD activity and induces Ca2+ mobilization and PLA2 activation in human platelets. Neomycin sulfate inhibits DNase I induced DNA degradation.
Formula:
C₂₃H₄₆N₆O₁₃·₃H₂SO₄·xH₂O Storage Temperature: Ambient |
MDL Number:
MFCD00150829 CAS Number: 1405-10-3 EINECS: 215-773-1 Merck Index: 14,6454 |
Specification Test Results
Potency | 600 µg/mg min. |
Loss on Drying | 8.0% max. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...