Polyethylene glycol 3,350 solution 50%

Supplier: Thermo Scientific Chemicals

Synonyms: PEG 3,350, PEG 3350

J61495-AE J61495-AK
CAAAJ61495-AEEA 105.23 CAD
CAAAJ61495-AE CAAAJ61495-AK
Polyethylene glycol 3,350 solution 50%
Polyethylene glycol 3,350

Viscous liquid


CAS No.: 25322-68-3 Molecular Formula: H(OCH2CH2)nOH Physical Form: Viscous liquid Appearance: Colorless MDL No.: MFCD00081839

Formula: H(OCH₂CH₂)nOH
MW: 3350 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00081839
CAS Number: 25322-68-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR