CHES, 0.5 M Buffer Solutions, Thermo Scientific

Supplier: Thermo Scientific Chemicals

J61492-AE J61492-AK J63670-AE J63670-AK
CAAAJ61492-AEEA 175.14 CAD
CAAAJ61492-AE CAAAJ61492-AK CAAAJ62333-AE CAAAJ62333-AK CAAAJ63419-AE CAAAJ63419-AK CAAAJ63670-AE CAAAJ63670-AK
CHES, 0.5 M Buffer Solutions, Thermo Scientific
Buffers Biological Buffers

2-(Cyclohexylamino)ethanesulfonic acid, CHES.


CAS No.: 103-47-9
MDL No.: MFCD00003835

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR