D-Amygdalin 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
J61472-03
J61472-06
J61472-14
CAAAJ61472-03EA
144.29
CAD
CAAAJ61472-03
CAAAJ61472-06
CAAAJ61472-14
D-Amygdalin 98%
D-Amygdalin
A benzylic glycoside that has been used as an antineoplastic agent
Purity: 98%
CAS No.: 29883-15-6
Molecular Formula: C20H27NO11
Formula Weight: 457.44
Physical Form: Powder
Appearance: White to off-white
Melting Point: 210-215°
Beilstein Reference: 66856
Optical Rotation: -38° (c=1.2 in water)
MDL No.: MFCD00006598
Applications: A benzylic glycoside that has been used as an antineoplastic agent
Formula:
C₂₀H₂₇NO₁₁ Melting Pt: 210…215 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006598 CAS Number: 29883-15-6 EINECS: 249-925-3 Merck Index: 14,597 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...