4-Aminopyridine 99+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
J61470-22
J61470-14
CAAAJ61470-22EA
279.66
CAD
CAAAJ61470-22
CAAAJ61470-14
4-Aminopyridine 99+%
4-Aminopyridine
A potassium channel blocker
Crystalline powder
A potassium channel blocker
Formula:
C₅H₆N₂ Boiling Pt: 273…274 °C Melting Pt: 157…161 °C Density: 1.26 Flash Pt: 164 °C(327 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006439 CAS Number: 504-24-5 EINECS: 207-987-9 UN: 2671 ADR: "6.1",II Merck Index: 14,3933 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...