MOPSO, 0.2 M Buffer Solution, pH 7.0, Thermo Scientific

Supplier: Thermo Scientific Chemicals

J61469-AE J61469-AK
CAAAJ61469-AEEA 170.01 CAD
CAAAJ61469-AE CAAAJ61469-AK
MOPSO, 0.2 M Buffer Solution, pH 7.0, Thermo Scientific
Buffers Biological Buffers

Liquid


CAS No.: 79803-73-9
Physical Form: Liquid
MDL No.: MFCD00070007

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR