L(+)-Histidine monohydrochloride monohydrate cell culture reagent
Supplier: Thermo Scientific Chemicals
J61465-09
J61465-22
J61465-36
CAAAJ61465-09EA
58.27
CAD
CAAAJ61465-09
CAAAJ61465-22
CAAAJ61465-36
CAAAJ61465-A1
L(+)-Histidine monohydrochloride monohydrate cell culture reagent
L(+)-Histidine monohydrochloride monohydrate
Powder
CAS No.: 5934-29-2
Molecular Formula: C6H9N3O2.HCl.H2O
Formula Weight: 209.63 (191.62anhy)
Physical Form: Powder
Appearance: White
Melting Point: ca 240° dec.
Beilstein Reference: 4168261
Merck Reference: 14,4720
Optical Rotation: +10° (c=5 in 5N HCl)
MDL No.: MFCD00151027
Formula:
C₆H₉N₃O₂·HCl·H₂O Melting Pt: ∼ 240 °C (dec.) Density: 1.485 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00151027 CAS Number: 5934-29-2 EINECS: 211-438-9 Merck Index: 14,4720 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...