β-Nicotinamide adenine dinucleotide reduced disodium salt trihydrate 98%

Supplier: Thermo Scientific Chemicals

J61461-03 J61461-06
CAAAJ61461-03EA 330.03 CAD
CAAAJ61461-03 CAAAJ61461-06
β-Nicotinamide adenine dinucleotide reduced disodium salt trihydrate 98%
beta-Nicotinamide adenine dinucleotide reduced disodium salt trihydrate

Powder


Purity: 98% Molecular Formula: C21H27N7Na2O14P2.3H2O Formula Weight: 763.46 (709.41anhy) Storage Temperature: -30°C to -10°C Physical Form: Powder Appearance: Off-white to pale yellow MDL No.: MFCD00150359

Formula: C₂₁H₂₇N₇Na₂O₁₄P₂·3H₂O
MW: 763.46 g/mol (709.41 anhy)
Storage Temperature: Freezer
MDL Number: MFCD00150359
CAS Number: 497934-76-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR