DNA (Deoxyribonucleic acid) (from salmon testes), Phenol extracted, denatured

Supplier: Thermo Scientific Chemicals

J61458-JEX J61458-JEX J61458-MA J61458-MA
CAAAJ61458-JEXEA 339.31 CAD
CAAAJ61458-JEX CAAAJ61458-MA AAJ61458-JEX AAJ61458-MA
DNA (Deoxyribonucleic acid) (from salmon testes), Phenol extracted, denatured
DNA (Deoxyribonucleic acid)

Phenol extracted liquid.


  • Store at Ambient temperatures

Storage Temperature: Ambient
MDL Number: MFCD00130922
CAS Number: 9007-49-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR