Furosemide ≥97%

Supplier: Thermo Scientific Chemicals
Danger

J61457-06 J61457-09 J61457-14
CAAAJ61457-06EA 248.39 CAD
CAAAJ61457-06 CAAAJ61457-09 CAAAJ61457-14
Furosemide ≥97%
Furosemide

Loop diuretic that inhibits the Na+/2Cl-/K+ (NKCC) cotransporter


Crystalline powder


Applications: Loop diuretic that inhibits the Na+/2Cl-/K+ (NKCC) cotransporter

Formula: C₁₂H₁₁ClN₂O₅S
Melting Pt: 206…209 °C
Storage Temperature: Ambient
MDL Number: MFCD00010549
CAS Number: 54-31-9
EINECS: 200-203-6
Merck Index: 14,4309

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR