Tulobuterol hydrochloride 98%

Supplier: Thermo Scientific Chemicals
Warning

J61448-03 J61448-06 J61448-MD
CAAAJ61448-03EA 416.95 CAD
CAAAJ61448-03 CAAAJ61448-06 AAJ61448-MD
Tulobuterol hydrochloride 98%
Tulobuterol hydrochloride

beta-2-adrenergic receptor agonist


Powder


Applications: beta-2-adrenergic receptor agonist

Formula: C₁₂H₁₈ClNO
Melting Pt: 159…163 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00214398
CAS Number: 56776-01-3
Merck Index: 14,9811

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR