Telmisartan 99%

Supplier: Thermo Scientific Chemicals

Synonyms: 4'-[[4-Methyl-6-(1-methyl-1H-benzimidazol-2-yl)-2-propyl-1H-benzimidazol-1-yl]methyl]biphenyl-2-carboxylic Acid, 4'-((1,7'-Dimethyl-2'-propyl-1H,3'H-[2,5'-bibenzo[d]imidazol]-3'-yl)methyl)-[1,1'-biphenyl]-2-carboxylic acid

J61441-03 J61441-MD
CAAAJ61441-03EA 509.95 CAD
CAAAJ61441-03 CAAAJ61441-MD
Telmisartan 99%
Telmisartan

An angiotensin II receptor antagonist


Powder


Applications: An angiotensin II receptor antagonist

Formula: C₃₃H₃₀N₄O₂
MW: 514.63 g/mol
Melting Pt: 261…263 °C
Storage Temperature: Ambient
MDL Number: MFCD00918125
CAS Number: 144701-48-4
Merck Index: 14,9129

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR