5-Hydroxyindol-3-ylacetic acid 98%

Supplier: Thermo Scientific Chemicals

J61435-03 J61435-ME
CAAAJ61435-03EA 264.81 CAD
CAAAJ61435-03 CAAAJ61435-ME
5-Hydroxyindol-3-ylacetic acid 98%
5-Hydroxyindol-3-ylacetic acid

Metabolite of melatonin


Crystalline powder


Metabolite of melatonin

Formula: C₁₀H₉NO₃
Melting Pt: 161…164 °C (dec.)
Storage Temperature: Freezer
MDL Number: MFCD00005639
CAS Number: 54-16-0
EINECS: 200-195-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR