5-Hydroxydecanoic acid sodium salt 98%

Supplier: Thermo Scientific Chemicals

J61434-MC J61434-ME
CAAAJ61434-MCEA 341.52 CAD
CAAAJ61434-MC CAAAJ61434-ME
5-Hydroxydecanoic acid sodium salt 98%
5-Hydroxydecanoic acid sodium salt

A potassium channel antagonist which blocks the postischemic effects of the potassium channel activator cromakalim


Powder


Applications: A potassium channel antagonist which blocks the postischemic effects of the potassium channel activator cromakalim

Formula: C₁₀H₁₉NaO₃
Storage Temperature: Refrigerator
MDL Number: MFCD00153808
CAS Number: 624-00-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR