Thymopentin ≥99%

Supplier: Thermo Scientific Chemicals

J61431-MB J61431-MC
CAAAJ61431-MBEA 313.79 CAD
CAAAJ61431-MB CAAAJ61431-MC
Thymopentin ≥99%
Thymopentin

Reduces endocrine responses


Powder


Applications: Reduces endocrine responses

Formula: C₃₀H₄₉N₉O₉
Storage Temperature: Freezer
MDL Number: MFCD00214200
CAS Number: 69558-55-0
Merck Index: 14,9403

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR