DL-Arginine hydrochloride

Supplier: Thermo Scientific Chemicals

J61420-06 J61420-14
CAAAJ61420-06EA 205.8 CAD
CAAAJ61420-06 CAAAJ61420-14
DL-Arginine hydrochloride
DL-Arginine hydrochloride

Substrate for nitric oxide synthase; induces insulin release by a nitric oxide-dependent mechanism


Powder


Applications: Substrate for nitric oxide synthase; induces insulin release by a nitric oxide-dependent mechanism

Formula: C₆H₁₄N₄O₂.HCl
Storage Temperature: Ambient
MDL Number: MFCD00064549
CAS Number: 32042-43-6
EINECS: 250-903-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR