Bezafibrate ≥98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-(4-(2-(4-Chlorobenzamido)ethyl)phenoxy)-2-methylpropanoic acid, 2-[4-[2-(4-Chlorobenzamido)ethyl]phenoxy]isobutyric Acid

J61412-03 J61412-06 J61412-ME
CAAAJ61412-03EA 98.15 CAD
CAAAJ61412-03 CAAAJ61412-06 CAAAJ61412-ME
Bezafibrate ≥98%
Bezafibrate

A peroxisome proliferator-activated receptor agonist and hypolipidemic agent


Powder


Applications: A peroxisome proliferator-activated receptor agonist and hypolipidemic agent

Formula: C₁₉H₂₀ClNO₄
MW: 361.82 g/mol
Melting Pt: 183…186 °C
Storage Temperature: Freezer
MDL Number: MFCD00078970
CAS Number: 41859-67-0
EINECS: 255-567-9
Merck Index: 14,1195

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR