2'-Deoxyinosine
Supplier: Thermo Scientific Chemicals
J61411-09
J61411-14
CAAAJ61411-09EA
319.59
CAD
CAAAJ61411-09
CAAAJ61411-14
CAAAJ61411-03
2'-Deoxyinosine
2'-Deoxyinosine
Crystalline powder
CAS No.: 890-38-0
Molecular Formula: C10H12N4O4
Formula Weight: 252.23
Storage Temperature: 2°C to 8°C
Physical Form: Crystalline powder
Appearance: White to off-white
Melting Point:
Formula:
C₁₀H₁₂N₄O₄ Melting Pt: >250 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00005762 CAS Number: 890-38-0 EINECS: 212-964-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...