MBS (N-Succinimidyl 3-maleimidobenzoate) 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Maleimidobenzoic acid N-hydroxysuccinimide ester, 3-Maleimidobenzoic Acid N-Succinimidyl Ester, MBS, m-Maleimidobenzoyl-N-hydroxysuccinimide ester, N-(3-Maleimidobenzoyloxy)succinimide

J61410-03 J61410-MD
CAAAJ61410-03EA 827.84 CAD
CAAAJ61410-03 CAAAJ61410-MD
MBS (N-Succinimidyl 3-maleimidobenzoate) 95%
MBS (N-Succinimidyl 3-maleimidobenzoate)

A heterobifunctional coupling reagent useful for forming enzyme immunoconjugates


Powder


Applications: A heterobifunctional coupling reagent useful for forming enzyme immunoconjugates

Formula: C₁₅H₁₀N₂O₆
Melting Pt: 175…177 °C
Storage Temperature: Freezer
MDL Number: MFCD00005514
CAS Number: 58626-38-3
EINECS: 261-368-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR