Clindamycin hydrochloride monohydrate ≥97%

Supplier: Thermo Scientific Chemicals
Warning

J61409-03 J61409-06
CAAAJ61409-03EA 490.65 CAD
CAAAJ61409-03 CAAAJ61409-06
Clindamycin hydrochloride monohydrate ≥97%
Clindamycin hydrochloride monohydrate

Antibiotic U-28508E, effective against Gram-positive bacteria


Powder


Applications: Antibiotic U-28508E, effective against Gram-positive bacteria

Formula: C₁₈H₃₃ClN₂O₅S
MW: 461.44 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06411091
CAS Number: 58207-19-5
EINECS: 244-398-6
Merck Index: 14,2356

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR