Carbonate-Buffered Saline (5X), pH 9.0, Thermo Scientific
Supplier: Thermo Scientific Chemicals
J61396-AK
J61396-AP
CAAAJ61396-AKEA
145.69
CAD
CAAAJ61396-AK
CAAAJ61396-AP
Carbonate-Buffered Saline (5X), pH 9.0, Thermo Scientific
Buffers
Cell Culture Buffers
Liquid
CAS No.: 497-19-8
Physical Form: Liquid
MDL No.: MFCD00003494
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...