Canrenoic acid potassium salt ≥98%

Supplier: Thermo Scientific Chemicals
Warning

J61394-03 J61394-06 J61394-14
CAAAJ61394-03EA 174.56 CAD
CAAAJ61394-03 CAAAJ61394-06 CAAAJ61394-14
Canrenoic acid potassium salt ≥98%
Canrenoic acid potassium salt

Powder


Purity: 98+% CAS No.: 2181-04-6 Molecular Formula: C22H30KO4 Formula Weight: 396.58 Physical Form: Powder Appearance: Pale yellow to brown MDL No.: MFCD05662375

Formula: C₂₂H₃₀KO₄
Storage Temperature: Ambient
MDL Number: MFCD05662375
CAS Number: 2181-04-6
EINECS: 218-554-9
Merck Index: 14,1750

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR