Phenethyl caffeate ≥99%

Supplier: Thermo Scientific Chemicals
Warning

J61386-03 J61386-MD
CAAAJ61386-03EA 1205.78 CAD
CAAAJ61386-03 CAAAJ61386-MD
Phenethyl caffeate ≥99%
Phenethyl caffeate

Inhibitor of protein tyrosine kinase, and is cytotoxic to cancer cell lines


Crystalline powder


Applications: Inhibitor of protein tyrosine kinase, and is cytotoxic to cancer cell lines

Formula: C₁₇H₁₆O₄
Melting Pt: 127…129 °C
Storage Temperature: Freezer
MDL Number: MFCD00866470
CAS Number: 104594-70-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR