Polyvinylpyrrolidone, M.W. ∼ 360,000 (K90)

Supplier: Thermo Scientific Chemicals

J61381-30 J61381-36 J61381-A1
CAAAJ61381-30EA 245.03 CAD
CAAAJ61381-30 CAAAJ61381-36 CAAAJ61381-A1
Polyvinylpyrrolidone, M.W. ∼ 360,000 (K90)
Polyvinylpyrrolidone

A cryoprotectant, also useful in plant cell culture research


Powder


A cryoprotectant, also useful in plant cell culture research

Formula: (C₆H₉NO)n
Storage Temperature: Ambient
MDL Number: MFCD01076626
CAS Number: 9003-39-8
Merck Index: 14,7697

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR