Sodium Phosphate, 0.2 M Buffer Solution, pH 8.5, Thermo Scientific
Supplier: Thermo Scientific Chemicals
J61372-AP
J61372-K2
CAAAJ61372-APEA
175.14
CAD
CAAAJ61372-AP
CAAAJ61372-K2
Sodium Phosphate, 0.2 M Buffer Solution, pH 8.5, Thermo Scientific
Buffers
Biological Buffers
Liquid
Physical Form: Liquid
MDL No.: MFCD00146206
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...