N-Methyl-D-aspartic acid ≥98%

Supplier: Thermo Scientific Chemicals

Synonyms: N-Methyl-D-asparaginic Acid, NMDA, Me-D-Asp-OH, (R)-2-(Methylamino)succinic acid

J61361-MC J61361-MF
CAAAJ61361-MCEA 461.9 CAD
CAAAJ61361-MC CAAAJ61361-ME CAAAJ61361-MF
N-Methyl-D-aspartic acid ≥98%
N-Methyl-D-aspartic acid

Powder


Purity: 98+% CAS No.: 6384-92-5 Molecular Formula: C5H9NO4 Formula Weight: 147.13 Storage Temperature: 2°C to 8°C Physical Form: Powder Sensitivity: Hygroscopic Appearance: White to off-white Melting Point: 187-192° Beilstein Reference: 1724431 Optical Rotation: -15° (c=2 in water) MDL No.: MFCD00004226

Formula: C₅H₉NO₄
MW: 147.13 g/mol
Melting Pt: 187…192 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004226
CAS Number: 6384-92-5
Merck Index: 14,6662

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR