7-Chlorokynurenic acid ≥99%

Supplier: Thermo Scientific Chemicals

Synonyms: 7-Chloro-4-hydroxyquinoline-2-carboxylic acid, 7-CTKA

J61359-MA J61359-MF
CAAAJ61359-MAEA 239.12 CAD
CAAAJ61359-MA CAAAJ61359-MF
7-Chlorokynurenic acid ≥99%
7-Chlorokynurenic acid

NMDA receptor antagonist at the glycine site


Powder


Applications: NMDA receptor antagonist at the glycine site

Formula: C₁₀H₆ClNO₃
MW: 223.62 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00069227
CAS Number: 18000-24-3
EINECS: 241-913-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR